CHIS5-02401

5-02402 5-02401
76223-060EA 302.97 USD
76223-060 76223-058
CHIS5-02401
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR