CHIS5-02395

5-02396 5-02395
76223-046EA 122.83 USD
76223-046 76223-048
CHIS5-02395
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR