CHIS5-02371

5-02372 5-02371
76223-000EA 193.25 USD
76223-000 76222-998
CHIS5-02371
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR