CHIS5-02365

5-02365 5-02366
76222-988EA 274.53 USD
76222-988 76222-986
CHIS5-02365
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR