CHIS5-02363

5-02364 5-02363
76225-762EA 68.78 USD
76225-762 76225-764
CHIS5-02363
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR