CHIS5-02339

5-02340 5-02339
76222-944EA 123.98 USD
76222-944 76222-942
CHIS5-02339
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR