CHIS5-02301

5-02302 5-02301
76222-874EA 49.13 USD
76222-874 76222-876
CHIS5-02301
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR