CHIS5-02293

5-02294 5-02293
76222-858EA 245.66 USD
76222-858 76222-860
CHIS5-02293
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR