CHIS5-02289

5-02289 5-02290
76225-742EA 73.7 USD
76225-742 76225-744
CHIS5-02289
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR