CHIS5-02243

5-02243 5-02244
76222-782EA 47.83 USD
76222-782 76222-784
CHIS5-02243
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR