CHIS5-02241

5-02242 5-02241
76222-780EA 360.29 USD
76222-780 76222-778
CHIS5-02241
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR