CHIS5-02237

5-02237 5-02238
76222-770EA 81.89 USD
76222-770 76222-772
CHIS5-02237
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR