CHIS5-02175

5-02175 5-02176
76222-680EA 573.2 USD
76222-680 76222-678
CHIS5-02175
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR