CHIS5-02169

5-02169 5-02170
76225-686EA 327.54 USD
76225-686 76225-688
CHIS5-02169
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR