CHIS5-02159

5-02159 5-02160
76222-672EA 471.66 USD
76222-672 76222-670
CHIS5-02159
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR