CHIS5-02151

5-02151 5-02152
76222-654EA 73.7 USD
76222-654 76222-656
CHIS5-02151
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR