CHIS5-02149

5-02150 5-02149
76222-650EA 181.79 USD
76222-650 76222-652
CHIS5-02149
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR