CHIS5-02143

5-02144 5-02143
76222-638EA 40.95 USD
76222-638 76222-640
CHIS5-02143
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR