CHIS5-02119

5-02119 5-02120
76222-292EA 147.39 USD
76222-292 76222-290
CHIS5-02119
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR