CHIS5-02115

5-02116 5-02115
76222-282EA 44.22 USD
76222-282 76222-284
CHIS5-02115
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR