CHIS5-02111

5-02111 5-02112
76222-276EA 196.52 USD
76222-276 76222-274
CHIS5-02111
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR