CHIS5-02137

5-02138 5-02137
76222-628EA 188.34 USD
76222-628 76222-626
CHIS5-02137
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR