CHIS5-02135

5-02136 5-02135
76225-666EA 36.03 USD
76225-666 76225-668
CHIS5-02135
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR