CHIS5-02131

5-02131 5-02132
76225-660EA 171.95 USD
76225-660 76225-658
CHIS5-02131
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR