CHIS5-02127

5-02127 5-02128
76225-652EA 497.86 USD
76225-652 76225-650
CHIS5-02127
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR