CHIS5-02125

5-02125 5-02126
76225-646EA 38.97 USD
76225-646 76225-648
CHIS5-02125
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR