You Searched For: Crimpers, Cappers and Decappers
Attach and remove seals and closures manually or with electric powered crimpers and decappers. Some models feature ergonomic, cushioned handles for maximum comfort. Adjustable sealing compression and autoclavable tapered jaws are some of the additional features also available.
Attach and remove seals and closures manually or with electric powered crimpers and decappers. Some models feature ergonomic, cushioned handles for maximum comfort. Adjustable sealing compression and autoclavable tapered jaws are some of the additional features also available.
81  results were found
SearchResultCount:"81"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR International
Description: VWR® Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles.
Supplier: VWR International
Description: Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
Catalog Number: (47743-758)
Supplier: Bel-Art Products, a Part of SP
Description: This convenient tool for opening microcentrifuge tubes is double-ended to work on all tube sizes.
Supplier: AZENTA US, INC
Description: An ideal range of manual devices to assist in tube picking and decapping.
New Product
Supplier: DWK Life Sciences (KIMBLE)
Description: EZ-Decappers™ Hand Operated Cappers and Decappers by MicroLiter are easy-to-use tools for crimping and decrimping chromatography vial closures.
Supplier: Thermo Scientific
Description: Use SureSTART™ level 3 manual vial crimpers for 20 mm Crimp vials to ensure that your crimp caps are sealed reliably every time.
Supplier: DWK Life Sciences (KIMBLE)
Description: Wheaton's High Performance Crimping Tool is designed with inter-changeable jaws, so that users only need one tool, and can purchase individually sized...
Supplier: Ace Glass
Description: This hand crimper is suitable for attaching aluminum seals.
Product available on GSA Advantage® Small Business Enterprise
Supplier: PERKINELMER U.S. LLC
Description: The electronic universal, precision control, power crimpers with adjustable settings are designed to provide multiple crimps on a single battery charg...
Supplier: AZENTA US, INC
Description: The semi-automated screw cap single tube decapper/recapper is the first-of-its-kind, single tube capper/de-capper in the industry. It provides a low-c...
Supplier: Ace Glass
Description: These decapper pliers are used for detaching aluminum seals only.
Small Business Enterprise
Supplier: AZENTA US, INC
Description: The Semi-automated screw cap decapper/recapper systems are compact, bench top units designed for efficient tube capping in labs with medium throughput...
Supplier: Thermo Scientific
Description: Use Thermo Scientific™ electronic hand-held vial crimpers and decrimpers to attach or remove crimp caps from vials, respectively.
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Description: A selection of ergonomically designed manual vial crimpers, vial decappers, and related accessories.
Supplier: SP Industries
Description: Easily remove end caps or base plugs.
Supplier: DWK Life Sciences (KIMBLE)
Description: These crimpers are used for flip cap seals and for aluminum seals sizes 16.5, 28.5, and 30 mm.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 81
no targeter for Bottom