BBL™ Brilliant Green Agar Plate, BD Biosciences
Supplier: BD Biosciences
BBL™
295963 295963
90004-612PK 122.35 USD
CA90004-612 90004-612
BBL™ Brilliant Green Agar Plate, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Brilliant green agar is a highly selective medium for the isolation of Salmonella other than S. Typhi from feces and other materials.
Brilliant green dye inhibits gram-positive bacteria and a majority of gram-negative bacilli. Phenol red serves as a pH indicator and yields a yellow color as a result of acid production in the fermentation of the lactose or sucrose in the medium.