L(-)-Malic acid disodium salt, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0521807310
76203-074EA
177.8
USD
76203-074
L(-)-Malic acid disodium salt, white powder
L(-)-Malic acid disodium salt
- Key Applications : Source of CO2 in the Calvin cycle; Used as a food additive
- Application Areas : Biochemistry
- Product Type : Biochemicals
- Chemical Class : Carbohydrates
- Presentation :White Powder
- Format : Powder
Specification Test Results
Identity Test | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...