Poly-L-lysine hydrobromide

Supplier: MP Biomedicals
0219454305
76204-002EA 83.61 USD
76204-002
Poly-L-lysine hydrobromide
Proteins and Peptides
Poly-L-lysine is a positively charged amino acid polymer. Poly-lysine binds to DNA, red cell membrane and any negatively charged protein. It is typically used as a coating substrate for culture dishes, slides, etc. It enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. When adsorbed to the culture surface, poly-lysine increases the number of positively charged sites available for cell binding.

  • Grade : Cell Culture Reagent, γ-irradiated
  • Suitability : suitable for cell culture
  • Key Applications : Attachment factor for tissue culture.
  • Application Areas : Cell Biology & Analysis
  • Product Type : Biochemicals
  • Presentation : Beige Powder
  • NOTES : There is approximately one HBr or HCl per lysine unit. The HBr/HCl allows it to be a crystalline solid and soluble in water. To remove the HBr/HCl, solubilize the product in a neutral buffer and dialyze away the salts.
  • Solubility : Soluble in water (50mg/mL) clear,colorless solution.
  • Sterility : γ-Irradiated
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR