Yeast Extract, Yellow-Tan Powder for Molecular Biology, MP Biomedicals

Supplier: MP Biomedicals
0219402783 0219402791
76204-082EA 86.6 USD
76204-082 76204-084
Yeast Extract, Yellow-Tan Powder for Molecular Biology, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Yeast extract powder is a vacuum dried autolyzed yeast extract produced from 100% Baker's yeast. It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.

  • Grade : Molecular Biology Reagent
  • Suitability : Suitable for alternative to beef extract
  • Key Applications : Bacteriological media component
  • Application Areas : Used as a bacteriological media component
  • Product Type : Proteins, Enzymes & Peptides
  • Presentation : Yellow-tan Powder
  • pH : 5 - 5.5 (aqueous soln.)(Lit.)
  • Foreign activity : Salmonella - Negative
  • Moisture content : <5%
  • NOTES : Mold Count ≤50/g, Standard Plate Count ≤50,000/g
  • Solubility : Solubility in water: - Clear (5 g/100 mL).
  • Mold Count: < 50/gram
  • Standard Plate Count: < 50,000/gram
  • Salmonella: Negative
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR