Novobiocin sodium salt, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0215595725
0215595701
76102-642EA
1003.38
USD
76102-642
76102-640
Novobiocin sodium salt, white powder
Novobiocin sodium salt
Novobiocin is an antibiotic produced by Streptomyces nieveus.
- Extinction Coefficient (E1%) : E1%= 600 (0.1 M NAOH)
- Optical Rotation : -38.0°(2.5 g/100 ml ethanol at 24°C for the free acid)
- UV/Visible Absorbance : λmax: 307 nm
- Residue on ignition : ≤12%
- Solubility : Freely soluble in wqter (10%-clear, colorless solution)
Formula:
C₃₁H₃₅N₂NaO₁₁ Melting Pt: 220 °C Density: 1,3448 g/ml Storage Temperature: Refrigerator |
MDL Number:
MFCD00066541 CAS Number: 1476-53-5 |
Specification Test Results
Identity Test | Passes |
Potency | ~900 µg/mg |
FTIR | Conforms to Standard |
Residue on Ignition |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...