Anti-Horse Serum Goat Polyclonal Antibody

Supplier: MP Biomedicals
0856002
IC0856002EA 211.12 USD
IC0856002
Anti-Horse Serum Goat Polyclonal Antibody
Antibodies
Product is the lyophilized powder of goat antiserum to horse serum and buffer salts.

Type: Primary 
Antigen: Horse Serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR