Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antithrombin (Human) ELISA Kit, BioVision, Inc.

Supplier: BioVision
E4526-100
A Sandwich ELISA kit for the quantitative measurement of antithrombin in biological fluids.

  • Easy, convenient and time-saving method to measure the level of antithrombin in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 31.25 - 2000 ng/ml
  • Sensitivity: < 18.75 ng/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%
  • This assay has high sensitivity and excellent specificity for detection of antithrombin. No significant cross-reactivity or interference between antithrombin and analogues was observed.

Antithrombin is most important serine protease inhibitor in plasma that regulates the blood coagulation cascade. Antithrombin inhibits thrombin, matriptase-3/TMPRSS7, as well as factors IXa, Xa and XIa. Its inhibitory activity is greatly enhanced in the presence of heparin. BioVision’s antithrombin ELISA kit is a sandwich ELISA assay for the quantitative measurement of antithrombin in Human serum, plasma and cell culture supernatants. The density of color is proportional to the amount of antithrombin captured from the samples.




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Antithrombin (Human) ELISA Kit 100 assays E4526-100 76236-020 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping