OxLDL (Mouse) ELISA Kit, BioVision

Supplier: BioVision
E4517-100
76237-738EA 852.27 USD
76237-738
OxLDL (Mouse) ELISA Kit, BioVision
A Sandwich ELISA kit for the quantitative measurement of OxLDL in biological fluids.

  • Easy, convenient and time-saving method to measure the level of OxLDL in Mouse serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 0.313 - 20 ng/ml
  • Sensitivity: < 0.188 ng/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%
  • This assay has high sensitivity and excellent specificity for detection of OxLDL. No significant cross-reactivity or interference between OxLDL and analogues was observed.

Low-density lipoprotein (LDL) are one of the five major groups of lipoprotein which transport all fat molecules around the body in the extracellular water. LDL delivers fat molecules to the cells and can drive the progression of atherosclerosis if they become oxidized within the walls of arteries. Since the Oxidized low-density lipoproteins (OxLDL) are more easily retained by the proteoglycans, a risk for cardiovascular disease when they invade the endothelium. BioVision’s OxLDL ELISA kit is a sandwich ELISA assay for the quantitative measurement of OxLDL in Mouse serum, plasma and cell culture supernatants. The density of color is proportional to the amount of OxLDL captured from the samples.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR