QuickDetect™ Anti-dsDNA Antibody (Human) ELISA Kit, BioVision

Supplier: BioVision
E4484-100
76236-056EA 601.26 USD
76236-056
QuickDetect™ Anti-dsDNA Antibody (Human) ELISA Kit, BioVision
A rapid sandwich ELISA kit for the quantitative measurement of anti-dsDNA Antibody in human samples.

  • Easy, convenient and time-saving method to measure the level of dsDNA in human serum, plasma, cell culture samples, biological fluid
  • Assay range: 0.1 IU/ml to 60 IU/ml
  • Sensitivity: 1.0 IU/ml
  • Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed

Anti-double stranded DNA (Anti-dsDNA) antibodies are a group of anti-nuclear antibodies (ANA) the target antigen of which is double stranded DNA. Blood tests such as enzyme-linked immunosorbent assay (ELISA) and immunofluorescence are routinely performed to detect anti-dsDNA antibodies in diagnostic laboratories. They are highly diagnostic of systemic lupus erythematosus (SLE) and are implicated in the pathogenesis of lupus nephritis. BioVision’s Anti-dsDNA ELISA kit is a sandwich ELISA assay for the qualitative measurement of Anti-dsDNA antibody in human serum, plasma and cell culture supernatants in 90 minutes.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR