QuickDetect™ Anti-ssDNA Antibody (Human) ELISA Kit, BioVision
Supplier: BioVision
E4483-100
76236-054EA 602.34 USD
76236-054
QuickDetect™ Anti-ssDNA Antibody (Human) ELISA Kit, BioVision
A rapid sandwich ELISA kit for the quantitative measurement of anti-ssDNA Antibody in human samples
  • Easy, convenient and time-saving method to measure the level of Anti-ssDNA in human Serum, plasma, cell culture samples, biological fluid.
  • Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed.
Antibodies against single-stranded DNA (anti-ssDNA) are mainly directed against its basic compound, which in the native DNA is masked inside the helical structure. Anti-ssDNA antibody (Ab) test is used for the differential diagnosis of new patients suspected of an inflammatory rheumatic disease the clinical sensitivity for systemic lupus erythematosus (SLE). The increase in anti-ssDNA Ab level appeared to be the best predictor of forthcoming increase in anti-dsDNA and SLE flare BioVision’s Anti-ssDNA ELISA kit is a sandwich ELISA assay for the qualitative measurement of Anti-ssDNA antibody in human serum, plasma and cell culture supernatants in 90 minutes.