Tetracycline hydrochloride 15 mg/ml in ethanol 70%

Supplier: TEKNOVA INC
Danger

T3325
100216-452EA 64.17 USD
100216-452
Tetracycline hydrochloride 15 mg/ml in ethanol 70%
Tetracycline hydrochloride

Bacteriostatic


Inhibits bacterial synthesis by binding to 30S ribosome. It is used as genetic marker in mammalian cell culture since no 30S ribosome (antibiotic wont accumulate).

Formula: C₂₂H₂₅ClN₂O₈
MW: 480.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078142
CAS Number: 64-75-5
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR