LigaTrap® Rat IgG Purification Kit, ImmunoReagents
LT-138KIT LT-138
75783-530EA 395.84 USD
75783-530 75783-518
LigaTrap® Rat IgG Purification Kit, ImmunoReagents
LigaTrap® Rat IgG Purification Kit uses LT technology to utilize novel, patented ligands, designed for antibody purification.
LigaTrap® Rat IgG Purification Resin binds an exceptionally broad range of recombinant, monoclonal and human or animal source polyclonal immunoglobulins.
Delivery information: Kit includes ten spin columns filled with LigaTrap® resin and buffers needed to capture species-specific antibodies.