Tetraethylammonium hydroxide
Supplier:
MilliporeSigma
Description:
Tetraethylammonium hydroxide (20% aqueous solution) for synthesis, 250ml^
Catalog Number:
(EM8.22149.9025)
Supplier:
MilliporeSigma
Description:
Tetraethylammonium hydroxide (20% aqueous solution) for synthesis, 25g^
Supplier:
TCI America
Description:
CAS Number: 77-98-5
MDL Number: MFCD00009024
Molecular Formula: C8H21NO
Molecular Weight: 147.26
Form: Clear Liquid
Color: Colorless
Supplier:
TCI America
Description:
CAS Number: 77-98-5
MDL Number: MFCD00009024
Molecular Formula: C8H21NO
Molecular Weight: 147.26
Form: Clear Liquid
Color: Colorless
Specific Grav...
Supplier:
Alfa Aesar
Description:
25% w/w in methanol,UN2924,Liquid,DANGER: FLAMMABLE, CORROSIVE, burns skin and eyes,250g,50g,FLAMMABLE LIQUID,CORROSIVE
Catalog Number:
(TCI0363-25ML)
Supplier:
TCI America
Description:
[Reagent for Ion-Pair Chromatography]
CAS Number: 77-98-5
MDL Number: MFCD00009024
Molecular Formula: C8H25NO
Molecular Weight: 147.26
Form: Clea...
Supplier:
Alfa Aesar
Description:
Grade: 20% aq. soln. , Melting Point C. Boiling Point C: NA. C8H21NO. 77-98-5. CORROSIVE AIR SENSITIVE
Supplier:
Alfa Aesar
Description:
35% w/w Aqueous Solution, Reagent Grade,UN3267,Liquid,DANGER: CORROSIVE, burns skin and eyes,100g,500g,CORROSIVE,
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
* Please note if your account is within the State of California two of these pieces of identification will be required.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
|
|||||||||