You Searched For: Acetylcholine bromide
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (TCA0083-025G)
Supplier: TCI America
Description: CAS Number: 66-23-9 MDL Number: MFCD00011814 Molecular Formula: C7H16BrNO2 Molecular Weight: 226.11 Purity/Analysis Method: 98.0% (T) Form: C...