Complete Control Systems, Agilent Technologies
Supplier: Agilent Technologies
217467 217461 217460 217564 217468 217468 240028 240028 217564 217461 217460 217467
ST217467EA 89.48 USD
CA95040-762 ST217467 CA99900-552 CA99900-574 99900-540 CAST217467 95040-806 CA99900-540 99900-574 99900-552 95040-762 CA95040-806
Complete Control Systems, Agilent Technologies
Protein Expression Protein Expression Systems
The Complete Control Inducible Mammalian Expression System is designed to provide the tightest control of expression available in mammalian cells. The system is based on a synthetic ecdysone-inducible receptor and a synthetic receptor recognition element that modulates expression of your gene of interest.
  • Dose-responsive inducible mammalian expression
  • Induction as high as 1700 fold
  • Novel IRES site allows expression of both receptor subunits from the same CMV promoter
  • Retroviral and plasmid gene delivery systems available
Caution: For Research Use Only. Not for Use in Diagnostic Procedures.