Sodium nitrate 99.0-100.5%, meets analytical specification of FCC, E251, Fluka™

Supplier: Honeywell Research Chemicals
Warning

13444-1KG
BJ-13444-1KGEA 123.49 USD
BJ-13444-1KG
Sodium nitrate 99.0-100.5%, meets analytical specification of FCC, E251, Fluka™
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 308 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.0 - 100.5 %
Loss on drying (105°C) max. 1 %
pH (5 %, 20°C) 5 to 7
Arsenic (As) max. 2 ppm
Calcium (Ca) max. 0.005 %
Copper (Cu) max. 0.001 %
Iron (Fe) max. 0.001 %
Mercury (Hg) max. 1 ppm
Potassium (K) max. 0.01 %
Magnesium (Mg) max. 0.002 %
Lead (Pb) max. 2 ppm
Zinc (Zn) max. 0.001 %
Heavy metals (as Pb) max. 0.001 %
Chloride (Cl) max. 0.001 %
Nitrite (NO₂) max. 10 ppm
Sulfate (SO₄) max. 0.01 %
Total halogen compounds (as Cl) max. 0.02 %
Residual solvents complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR