Dimethylzinc ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

97-5061-25G 97-5061-100G
100205-808EA 4247.57 USD
100205-808 100209-470
Dimethylzinc ≥99%
Dimethylzinc

Metal Alkyls, Volatile Precursors for CVD

Formula: C₂H₆Zn
Boiling Pt: 46 °C
Melting Pt: –42 °C
Density: 1,386 (20°)
Flash Pt: –1 °F
MDL Number: MFCD00014854
CAS Number: 544-97-8
UN: 3394
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR