Mercury(II) perchlorate trihydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-8042-50G
93-8042-10G
100204-436EA
45.29
USD
100204-436
100209-220
Mercury(II) perchlorate trihydrate ≥99%
Mercury (II) perchlorate trihydrate
Formula:
Cl₂HgO₈·3H₂O MW: 453.529 g/mol Density: ∼4 |
MDL Number:
MFCD00149735 CAS Number: 73491-34-6 UN: 3087 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...