Tantalum(V) ethanolate ≥99.99% (Ta basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tantalum(V) ethoxide

93-7303-10G 93-7303-250G 93-7303-50G
100207-314EA 1635 USD
100207-314 100200-604 100207-150
Tantalum(V) ethanolate ≥99.99% (Ta basis), PURATREM
Tantalum(V) ethanolate

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₁₀H₂₅O₅Ta
MW: 406.25 g/mol
Boiling Pt: 145 °C (0.1 mmHg)
Melting Pt: 21 °C
Density: 1,56
Flash Pt: 87 °F
MDL Number: MFCD00049785
CAS Number: 6074-84-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR