Ytterbium(III) perchlorate 50% in aqueous solution ≥99.9% (Yb basis)

Warning

93-7010-2G
100207-144EA 52.37 USD
100207-144
Ytterbium(III) perchlorate 50% in aqueous solution ≥99.9% (Yb basis)
Ytterbium (III) perchlorate
Formula: Cl₃O₁₂Yb
MW: 471.392 g/mol
MDL Number: MFCD00049612
CAS Number: 13498-08-3
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR