Dysprosium(III) carbonate tetrahydrate ≥99.9% (Dy basis)
Supplier: STREM CHEMICALS INC MS
93-6627-25G
93-6627-100G
100210-526EA
324.12
USD
100210-526
100199-650
Dysprosium(III) carbonate tetrahydrate ≥99.9% (Dy basis)
Dysprosium (III) carbonate tetrahydrate
Formula:
C₃Dy₂O₉·4H₂O |
MDL Number:
MFCD00150229 CAS Number: 38245-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...