Dysprosium trinitrate hexahydrate ≥99.9% (Dy basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Dysprosium(III) nitrate hexahydrate
93-6613-100G
93-6613-25G
100198-436EA
774.22
USD
100198-436
100202-256
Dysprosium trinitrate hexahydrate ≥99.9% (Dy basis)
Dysprosium trinitrate hexahydrate
Formula:
DyN₃O₉·6H₂O MW: 456.606 g/mol |
MDL Number:
MFCD01311712 CAS Number: 35725-30-5 UN: 1477 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...