Gadolinium(III) sulfate octahydrate ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6407-100G 93-6407-25G
100203-554EA 287.32 USD
100203-554 100204-404
Gadolinium(III) sulfate octahydrate ≥99.9% (Gd basis)
Gadolinium (III) sulphate octahydrate
Formula: Gd₂O₁₂S₃·8H₂O
MW: 746.81 g/mol
Density: 3,010
MDL Number: MFCD00149734
CAS Number: 13450-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR